missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Ro 15-4513

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
High affinity benzodiazepine ligand. Ki values are 3.1 and 5.3 nM for diazepam-insensitive (DI) and diazepam-sensitive (DS) benzodiazepine receptors respectively. Acts as partial inverse agonist at recombinant DS α1-, α2-, α3- and α5-GABAA receptors. Displays partial agonism at DI α4- and α6-GABAA receptors. Antagonizes several behavioral and neurochemical effects of ethanol. Proconvulsant and anxiogenic.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 326.31 |
| Purity | >98% |
| Molecular Weight (g/mol) | 326.316 |
| Chemical Name or Material | Ro 15-4513 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction