missing translation for 'onlineSavingsMsg'
Learn More

Ro 10-5824 dihydrochloride, MedChemExpress

Product Code. 16476067
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476067 5 mg 5mg
16403348 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16476067 Supplier Medchem Express Supplier No. HY101384A
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Ro 10-5824 dihydrochloride is a selective dopamine D4 receptor partial agonist, with Ki of 5.2 nM.

Ro 10-5824 dihydrochloride is a selective dopamine D4 receptor partial agonist, with Ki of 5.2 nM.

Specifications

Chemical Name or Material Ro 10-5824 dihydrochloride
Physical Form Solid
CAS 189744-94-3
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C17H22Cl2N4
Solubility Information H2O : 100 mg/mL (283.05 mM; Need ultrasonic) ∣DMSO : 8.33 mg/mL (23.58 mM; Need ultrasonic)
SMILES NC1=NC(C)=NC=C1CN2CC=C(C3=CC=CC=C3)CC2.[H]Cl.[H]Cl
Molecular Weight (g/mol) 353.29
Formula Weight 353.29
Percent Purity 99.93%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.