missing translation for 'onlineSavingsMsg'
Learn More

RNPS1 Antibody (2G11), Novus Biologicals™

Product Code. 18394519 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394519 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18394519 Supplier Novus Biologicals Supplier No. H00010921M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RNPS1 Monoclonal antibody specifically detects RNPS1 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNPS1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2G11
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_006702
Gene Alias E5.1, LDC2, MGC117332, RNA binding protein S1, serine-rich domain, RNA-binding protein S1, serine-rich domain, RNA-binding protein with serine-rich domain 1, SR protein, SR-related protein LDC2
Host Species Mouse
Immunogen RNPS1 (NP_006702, 158 a.a. ~ 239 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PKPTKVHIGRLTRNVTKDHIMEIFSTYGKIKMIDMPVERMHPHLSKGYAYVEFENPDEAEKALKHMDGGQIDGQEITATAVL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10921
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.