missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNPEPL1 Antibody (4D12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59372
This item is not returnable.
View return policy
Description
RNPEPL1 Monoclonal specifically detects RNPEPL1 in Non-species specific samples. It is validated for Western Blot.
Specifications
| RNPEPL1 | |
| Monoclonal | |
| Unconjugated | |
| PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide | |
| RNPEPL1 | |
| Synthetic N-terminal arginylated peptide conjugated to KLH | |
| 100 μg | |
| Primary | |
| Specific for N-terminal arginine, next to both glutamic acid and aspartic acid. Does not detect internal arginine. | |
| Store at -20C. | |
| IgG1 |
| Western Blot | |
| 4D12 | |
| Western Blot 1:100 - 1:2000 | |
| argininyl aminopeptidase-like 1, arginyl aminopeptidase (aminopeptidase B)-like 1, arginyl aminopeptidase-like 1, EC 3.4.11.-, FLJ10806, FLJ26675, MGC99544, RNPEP-like 1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 57140 | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction