missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNPEPL1 Antibody (4D12), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59372G
This item is not returnable.
View return policy
Description
RNPEPL1 Monoclonal specifically detects RNPEPL1 in Non-species specific samples. It is validated for Western Blot.
Specifications
| RNPEPL1 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RNPEPL1 | |
| Synthetic N-terminal arginylated peptide conjugated to KLH | |
| 100 μL | |
| 57140 | |
| Non-species specific | |
| Purified |
| Western Blot | |
| 4D12 | |
| Western Blot | |
| argininyl aminopeptidase-like 1, arginyl aminopeptidase (aminopeptidase B)-like 1, arginyl aminopeptidase-like 1, EC 3.4.11.-, FLJ10806, FLJ26675, MGC99544, RNPEP-like 1 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Specific for N-terminal arginine, next to both glutamic acid and aspartic acid. Does not detect internal arginine. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction