missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNPEPL1 Antibody (4A9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59371-25ug
This item is not returnable.
View return policy
Description
RNPEPL1 Monoclonal specifically detects RNPEPL1 in Non-species specific samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| RNPEPL1 | |
| 4A9 | |
| Western Blot 1:100 - 1:2000 | |
| argininyl aminopeptidase-like 1, arginyl aminopeptidase (aminopeptidase B)-like 1, arginyl aminopeptidase-like 1, EC 3.4.11.-, FLJ10806, FLJ26675, MGC99544, RNPEP-like 1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 57140 | |
| Store at -20°C. | |
| IgG1 |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 50% glycerol with 0.09% Sodium Azide | |
| RNPEPL1 | |
| Synthetic N-terminal arginylated peptide conjugated to KLH | |
| 25 μg | |
| Primary | |
| Specific for N-terminal arginine next to glutamic acid. Does not detect N-terminal arginine next to aspartic acid or internal arginine residues. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction