missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNPEP Monoclonal Antibody (OTI3B5)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525783
This item is not returnable.
View return policy
Description
RNPEP Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Exopeptidase which selectively removes arginine and/or lysine residues from the N-terminus of several peptide substrates including Arg(0)-Leu-enkephalin, Arg(0)-Met-enkephalin and Arg(-1)-Lys(0)-somatostatin-14. Can hydrolyze leukotriene A4 (LTA-4) into leukotriene B4 (LTB-4).
Specifications
| RNPEP | |
| Monoclonal | |
| 0.77 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H4A4 | |
| Rnpep | |
| Full length human recombinant protein of RNPEP produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI3B5 | |
| Unconjugated | |
| Rnpep | |
| aminopeptidase B; APB; AP-B; Arginine aminopeptidase; arginyl aminopeptidase; arginyl aminopeptidase (aminopeptidase B); cytosol aminopeptidase IV; Rnpep; zgc:100971 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6051 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction