missing translation for 'onlineSavingsMsg'
Learn More

RNPC1 Antibody, Novus Biologicals™

Product Code. 18380989 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18380989 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18380989 Supplier Novus Biologicals Supplier No. H00055544B02P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

RNPC1 Polyclonal antibody specifically detects RNPC1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNPC1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_059965.2
Gene Alias CLL-associated antigen KW-5, HSRNASEBRRM) containing 1, RNA binding motif protein 38, RNA-binding motif protein 38, RNA-binding protein 38, RNA-binding region containing protein 1, RNA-binding region-containing protein 1, SEB4, seb4B, ssDNA binding protein SEB4, ssDNA-binding protein SEB4
Host Species Mouse
Immunogen RBM38 (NP_059965.2, 1 a.a. - 239 a.a.) full-length human protein. MLLQPAPCAPSAGFPRPLAAPGAMHGSQKDTTFTKIFVGGLPYHTTDASLRKYFEGFGDIEEAVVITDRQTGKSRGYGFVTMADRAAAERACKDPNPIIDGRKANVNLAYLGAKPRSLQTGFAIGVQQLHPTLIQRTYGLTPHYIYPPAIVQPSVVIPAAPVPSLSSPYIEYTPASPAYAQYPPATYDQYPYAASPATAASFVGYSYPAAVPQALSAAAPAGTTFVQYQAPQLQPDRMQ
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55544
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.