missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNMTL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554671
This item is not returnable.
View return policy
Description
Immunogen sequence: ILLEGRRLIS DALKAGAVPK MFFFSRLEYL KELPVDKLKG VSLIKVKFED IKDWSDLVTP QGIMGIFAKP DHVKMTYPKT QLQHSLPLLL ICDNLRDPGN Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 95%.
The RNA methyltransferase like 1 gene (RNMTL1) is one of thirteen newly discovered genes within a 116 Kb segment of the chromosome 17p13.3, which suffers from a high frequent loss of heterozygosity in human hepatocellular carcinoma. Its amino acid sequence was identified a high conserved region with methylases from various microorganisms, suggesting to have function to transfer methyl group to rRNA molecule.
Specifications
| RNMTL1 | |
| Polyclonal | |
| Unconjugated | |
| MRM3 | |
| 16S rRNA (guanosine(1370)-2'-O)-methyltransferase; 16S rRNA [Gm1370] 2'-O-methyltransferase; 4833420N02Rik; AI255153; HC90; mitochondrial rRNA methyltransferase 3; MRM3; putative RNA methyltransferase; RGD1309077; RMTL1; RNA methyltransferase like 1; RNA methyltransferase-like protein 1; RNMTL1; rRNA methyltransferase 3, mitochondrial | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55178 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HC36 | |
| MRM3 | |
| Recombinant protein corresponding to Human RNMTL1. Recombinant protein control fragment (Product #RP-93453). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction