missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNMTL1 Mouse anti-Human, Polyclonal, Abnova™
Mouse polyclonal antibody raised against a full-length human RNMTL1 protein.
Brand: Abnova H00055178-B01P.50ug
This item is not returnable.
View return policy
Specifications
| RNMTL1 | |
| Polyclonal | |
| RNA methyltransferase like 1 | |
| RNMTL1 | |
| FLJ10581, HC90 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 55178 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_018146 | |
| MRM3 | |
| RNMTL1 (NP_060616, 1 a.a. ∽ 420 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction