Learn More
RNMT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13743-1-AP-20UL
 View more versions of this product
Product Code. 16882114
  • £135.00 / 20µL
Description
RNMT belongs to the mRNA cap methyltransferase family. RNMT is a mRNA-capping methyltransferase that methylates the N7 position of the added guanosine to the 5′-cap structure of mRNAs. It binds RNA containing 5′-terminal GpppC.
Specifications
Specifications
RNMT
Polyclonal
Unconjugated
RNMT
hcm1p, hCMT1, hCMT1c, hMet, KIAA0398, MET, mRNA cap methyltransferase, RG7MT1, RNMT
Rabbit
Antigen Affinity Chromatography
RUO
291534, 67897, 8731
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O43148, Q5U2U7, Q9D0L8
Rnmt
RNMT Fusion Protein Ag4682
20 μL
Primary
Mouse, Rat, Human