missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNMT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13743-1-AP-150UL
This item is not returnable.
View return policy
Description
RNMT belongs to the mRNA cap methyltransferase family. RNMT is a mRNA-capping methyltransferase that methylates the N7 position of the added guanosine to the 5′-cap structure of mRNAs. It binds RNA containing 5′-terminal GpppC.Specifications
| RNMT | |
| Polyclonal | |
| Unconjugated | |
| RNMT | |
| hcm1p, hCMT1, hCMT1c, hMet, KIAA0398, MET, mRNA cap methyltransferase, RG7MT1, RNMT | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 291534, 67897, 8731 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43148, Q5U2U7, Q9D0L8 | |
| Rnmt | |
| RNMT Fusion Protein Ag4682 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction