Learn More
Invitrogen™ RNMT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113700
Description
Immunogen sequence: ETEDVPKDKS STGDGTQNKR KIALEDVPEK QKNLEEGHSS TVAAHYNELQ For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 79%; Human: 100%; Rabbit: 79%.
RNMT belongs to the mRNA cap methyltransferase family. RNMT is a mRNA-capping methyltransferase that methylates the N7 position of the added guanosine to the 5'-cap structure of mRNAs. It binds RNA containing 5'-terminal GpppC.
Specifications
| RNMT | |
| Polyclonal | |
| Unconjugated | |
| Rnmt | |
| 2610002P10Rik; AI848273; cm1p; CMT1; CMT1c; hcm1p; hCMT1; hMet; Kiaa0398; MET; mKIAA0398; mRNA (guanine-7-)methyltransferase; mRNA (guanine-N(7)-)-methyltransferase; mRNA cap guanine-N7 methyltransferase; mRNA cap methyltransferase; RG7MT1; RNA (guanine-7-) methyltransferase; RNA guanine-7 methyltransferase; RNMT | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 8731 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O43148 | |
| Rnmt | |
| Synthetic peptide directed towards the N terminal region of human RNMT (aa 108-157). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.