missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNMT, Mouse anti-Human, Clone: 3H3-1D12, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00008731-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MANSAKAEEYEKMSLEQAKASVNSETESSFNINENTTASGTGLSEKTSVCRQVDIARKRKEFEDDLVKESSSCGKDTPSKKRKLDPEIVPEEKDCGDAEGNSKKRKRETEDVPKDKSSTGDGTQNKRKIALEDVPEKQKNLEEGHSSTVAAHYNELQEVGLEKRSQSRIFYLRNFNNWMKSVLIGEFLEKVRQKKKRDITVLDLGCGKGGDLLKWKKGRINKLVCTDIADVSVKQCQQRYEDMKNRRDSEYIFSAEFITADSSKELLIDKFRDPQMCFDICSCQFVCHYSFESYEQADMMLRNACERLSPGGYFIGTTPNSFELIRRLEASETESFGNEIYTVKFQKKGDYPLFGCKYDFNLEGVVDVPEFLVYFPLLNEMAKKYNMKLVYKKTFLEFYEEKIKNNENKMLLKRMQALEPYPANESSKLVSEKVDDYEHAAKYMKNSQVRLPLGTLSKSEWEATSIYLVFAFEKQQSpecifications
| RNMT | |
| Monoclonal | |
| Unconjugated | |
| RNA (guanine-7-) methyltransferase | |
| DKFZp686H1252/KIAA0398/MET/RG7MT1/hCMT1c | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 3H3-1D12 | |
| In 1x PBS, pH 7.4 | |
| BC036798 | |
| RNMT | |
| RNMT (AAH36798, 1 a.a. ∼ 476 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 8731 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction