Learn More
Invitrogen™ RNH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579931
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human placenta tissue, human Hela whole cell, human SK-OV-3 whole cell, human Jurkat whole cell, human U-87MG whole cell, monkey COS-7 whole cell, human SW620 whole cell, human Caco-2 whole cell, rat brain tissue, rat testis tissue, rat lung tissue, mouse brain tissue, mouse testis tissue, mouse lung tissue, mouse HEPA1-6 whole cell. IHC: human intestinal cancer tissue, human intestinal cancer tissue, human lung cancer tissue, human lung cancer tissue. Flow: A549 cell, A549 cell.
RNH1 is the inhibitor of pancreatic RNase and angiogenin. RNH1 may also function in the modulation of cellular activities.
Specifications
| RNH1 | |
| Polyclonal | |
| Unconjugated | |
| RNH1 | |
| AW546468; C80305; EGK_05816; hypothetical protein LOC517087; im:7039360; LOC720791; Placental ribonuclease inhibitor; placental RNase inhibitor; PRI; RAI; Ribonuclease inhibitor; ribonuclease/angiogenin inhibitor 1; RNasIN; RNH; Rnh1; si:ch211-213a13.3; testicular tissue protein Li 164; uncharacterized protein rnh1; wu:fd55g05 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100360501, 107702, 6050 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| P13489, P29315, Q91VI7 | |
| RNH1 | |
| A synthetic peptide corresponding to a sequence at the N-terminus of human RNH1 (2-37aa SLDIQSLDIQCEELSDARWAELLPLLQQCQVVRLDD). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.