missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNH1 Monoclonal Antibody (OTI1B7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525307
This item is not returnable.
View return policy
Description
RNH1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
RNH1 is the inhibitor of pancreatic RNase and angiogenin. RNH1 may also function in the modulation of cellular activities.
Specifications
| RNH1 | |
| Monoclonal | |
| 1.00 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P13489 | |
| RNH1 | |
| Full length human recombinant protein of RNH1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI1B7 | |
| Unconjugated | |
| RNH1 | |
| AW546468; C80305; EGK_05816; hypothetical protein LOC517087; im:7039360; LOC720791; Placental ribonuclease inhibitor; placental RNase inhibitor; PRI; RAI; Ribonuclease inhibitor; ribonuclease/angiogenin inhibitor 1; RNasIN; RNH; Rnh1; si:ch211-213a13.3; testicular tissue protein Li 164; uncharacterized protein rnh1; wu:fd55g05 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6050 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction