missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNFT2 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06145PCP
This item is not returnable.
View return policy
Description
RNFT2 Polyclonal antibody specifically detects RNFT2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| RNFT2 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| FLJ14627, RING finger and transmembrane domain-containing protein 2, ring finger protein, transmembrane 2, TMEM118, transmembrane protein 118 | |
| Produced in rabbits immunized with E. coli-derived Human RNFT2 fragment. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 84900 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction