Learn More
Invitrogen™ RNF8 Recombinant Rabbit Monoclonal Antibody (9V0X5)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558050
Description
Immunogen sequence: MGEPGFFVTG DRAGGRSWCL RRVGMSAGWL LLEDGCEVTV GRGFGVTYQL VSKICPLMIS RNHCVLKQNP EGQWTIMDNK SLNGVWLNRA RLEPLRVYSI HQGDYIQLGV PLENKENAEY EYEVTEEDWE TIYPCLSPKN DQMIEKNKEL RTKRKFSLDE LAGPGAEGPS NLKSKINKVS CESGQPVKSQ GKGEVASTPS DNLDPKLTAL EPSKTTGAPI YPGFPKVTEV HHEQKASNSS ASQRSLQMFK VTMSRILRLK.
The protein encoded by this gene contains a RING finger motif and a FHA domain. This protein has been shown to interact with several class II ubiquitin-conjugating enzymes (E2), including UBE2E1/UBCH6, UBE2E2, and UBE2E3, and may act as an ubiquitin ligase (E3) in the ubiquitination of certain nuclear proteins. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
| RNF8 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| O76064, Q4KLN8, Q8VC56 | |
| RNF8 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human RNF8 (NP_003949.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 9V0X5 | |
| Unconjugated | |
| RNF8 | |
| 3830404E21Rik; ActA binding protein 2; actA-interacting protein 37; AIP37; C3HC4-type zinc finger protein; E3 ubiquitin-protein ligase RNF8; FLJ12013; hRNF8; KIAA0646; LaXp180; OTTHUMP00000039684; ring finger protein (C3HC4 type) 8; ring finger protein 8; ring finger protein 8, E3 ubiquitin protein ligase; RING-type E3 ubiquitin transferase RNF8; RNF8; UBC13/UEV-interacting ring finger protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 361815, 58230, 9025 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.