missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNF8 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12809112
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12809112 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12809112 Supplier Invitrogen™ Supplier No. PA520961

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is human lung tissue lysate. PA5-20961 can be used with blocking peptide PEP-1075.

The protein encoded by this gene contains a RING finger motif and a FHA domain. This protein has been shown to interact with several class II ubiquitin-conjugating enzymes (E2), including UBE2E1/UBCH6, UBE2E2, and UBE2E3, and may act as an ubiquitin ligase (E3) in the ubiquitination of certain nuclear proteins. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF8
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene RNF8
Gene Accession No. O76064, Q4KLN8, Q8VC56
Gene Alias 3830404E21Rik; ActA binding protein 2; actA-interacting protein 37; AIP37; C3HC4-type zinc finger protein; E3 ubiquitin-protein ligase RNF8; FLJ12013; hRNF8; KIAA0646; LaXp180; OTTHUMP00000039684; ring finger protein (C3HC4 type) 8; ring finger protein 8; ring finger protein 8, E3 ubiquitin protein ligase; RING-type E3 ubiquitin transferase RNF8; RNF8; UBC13/UEV-interacting ring finger protein
Gene Symbols RNF8
Host Species Rabbit
Immunogen A 14 amino acid peptide near the carboxy terminus of human RNF8.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361815, 58230, 9025
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.