missing translation for 'onlineSavingsMsg'
Learn More

RNF8 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18419532 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419532 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419532 Supplier Novus Biologicals Supplier No. NBP177166AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RNF8 Polyclonal antibody specifically detects RNF8 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF8
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias C3HC4-type zinc finger protein, EC 6.3.2, EC 6.3.2.-, FLJ12013, KIAA0646UBC13/UEV-interacting ring finger protein, ring finger protein (C3HC4 type) 8, ring finger protein 8E3 ubiquitin-protein ligase RNF8
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human RNF8. The immunogen is located within amino acids 330 - 380 of RNF8. Amino Acid Squence: MEELNRSKKDFEAI
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 9025
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.