missing translation for 'onlineSavingsMsg'
Learn More

RNF70 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18328570 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328570 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18328570 Supplier Novus Biologicals Supplier No. NBP2856510.1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RNF70 Polyclonal specifically detects RNF70 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF70
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 2% Sucrose
Gene Alias EC 6.3.2, EC 6.3.2.-, FLJ11830, praja 1, praja ring finger 1, PRAJA1, RING finger protein 70, RNF70E3 ubiquitin-protein ligase Praja-1
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the middle region of human RNF70
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 64219
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.