missing translation for 'onlineSavingsMsg'
Learn More

RNF70 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18417937 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417937 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18417937 Supplier Novus Biologicals Supplier No. NBP281998MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RNF70 Polyclonal antibody specifically detects RNF70 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF70
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EC 6.3.2, EC 6.3.2.-, FLJ11830, praja 1, praja ring finger 1, PRAJA1, RING finger protein 70, RNF70E3 ubiquitin-protein ligase Praja-1
Host Species Rabbit
Immunogen RNF70 antibody was raised against a 19 amino acid peptide near the amino terminus of human RNF70. The immunogen is located within amino acids 50 - 100 of RNF70. Amino Acid Squence: QTTKRSRSPFSTTRRSWDD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 64219
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.