missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF7 Recombinant Rabbit Monoclonal Antibody (24GB5200)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557230
This item is not returnable.
View return policy
Description
RNF7 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The protein encoded by this gene is a highly conserved ring finger protein. It is an essential subunit of SKP1-cullin/CDC53-F box protein ubiquitin ligases, which are a part of the protein degradation machinery important for cell cycle progression and signal transduction. This protein interacts with, and is a substrate of, casein kinase II (CSNK2A1/CKII). The phosphorylation of this protein by CSNK2A1 has been shown to promote the degradation of IkappaBalpha (CHUK/IKK-alpha/IKBKA) and p27Kip1(CDKN1B). Alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
| RNF7 | |
| Recombinant Monoclonal | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UBF6, Q9WTZ1 | |
| RNF7 | |
| A synthesized peptide derived from human RNF7 (aa 80-113). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB5200 | |
| Unconjugated | |
| RNF7 | |
| CKBBP1; CKII beta-binding protein 1; OTTHUMP00000180645; rbx2; Regulator of cullins 2; ring finger protein 7; RING-box protein 2; RNF7; ROC2; SAG; Sensitive to apoptosis gene protein; sensitive to apoptosis, zinc RING finger protein SAG, regulator of cullins 2; zinc RING finger protein SAG | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19823, 300948, 9616 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction