missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF6 Antibody (6D5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00006049-M02
This item is not returnable.
View return policy
Description
RNF6 Monoclonal antibody specifically detects RNF6 in Human samples. It is validated for Western Blot, ELISA
Specifications
| RNF6 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| DKFZp686P0776, E3 ubiquitin-protein ligase RNF6, EC 6.3.2.-, ring finger protein (C3H2C3 type) 6, RING-H2 protein RNF-6, SPG2 | |
| RNF6 (NP_005968, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MNQSRSRSDGGSEETLPQDHNHHENERRWQQERLHREEAYYQFINELNDEDYRLMRDHNLLGTPGEITSEELQQRLDGVKEQLASQPDLRDGTNYRDSEV | |
| 0.1 mg | |
| Zinc Finger | |
| 6049 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA | |
| 6D5 | |
| Western Blot, ELISA | |
| NP_005968 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction