missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110310
This item is not returnable.
View return policy
Description
Immunogen sequence: MAAAEEEDGG PEGPNRERGG AGATFECNIC LETAREAVVS VCGHLYCWPC LHQWLETRPE RQECPVCKAG ISREKVVPLY GRGSQKPQDP RLKTPPRPQG QRPAPESRGG FQPFGDTGGF.
The protein encoded by this gene contains a RING finger, which is a motif known to be involved in protein-protein interactions. This protein is a membrane-bound ubiquitin ligase. It can regulate cell motility by targeting paxillin ubiquitination and altering the distribution and localization of paxillin in cytoplasm and cell focal adhesions.
Specifications
| RNF5 | |
| Polyclonal | |
| Unconjugated | |
| RNF5 | |
| 2410131O05Rik; AA407576; E3 ubiquitin-protein ligase RNF5; G16; HsRma1; ing finger protein 5 pseudogene; NG2; Protein G16; ram1 homolog; RING finger protein 5; ring finger protein 5, E3 ubiquitin protein ligase; RING5; RING-type E3 ubiquitin transferase RNF5; RMA1; RNF5 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 407784, 54197, 6048 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.03 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O35445, Q5M807, Q99942 | |
| RNF5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human RNF5 (NP_008844.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction