missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566741
This item is not returnable.
View return policy
Description
Immunogen sequence: NAHEPFRRGTG VDLGQGHPAS SW Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.
The protein encoded by this gene contains a RING finger, which is a motif known to be involved in protein-protein interactions. This protein is a membrane-bound ubiquitin ligase. It can regulate cell motility by targeting paxillin ubiquitination and altering the distribution and localization of paxillin in cytoplasm and cell focal adhesions.
Specifications
| RNF5 | |
| Polyclonal | |
| Unconjugated | |
| RNF5 | |
| 2410131O05Rik; AA407576; E3 ubiquitin-protein ligase RNF5; G16; HsRma1; ing finger protein 5 pseudogene; NG2; Protein G16; ram1 homolog; RING finger protein 5; ring finger protein 5, E3 ubiquitin protein ligase; RING5; RING-type E3 ubiquitin transferase RNF5; RMA1; RNF5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6048 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99942 | |
| RNF5 | |
| Recombinant Human RNF5. Recombinant protein control fragment (Product #RP-107397). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction