missing translation for 'onlineSavingsMsg'
Learn More

RNF40 Rabbit anti-Human, Mouse, Clone: 8I7R5, Novus Biologicals™

Product Code. 18353062 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18353062 100 μg 100µL
18319476 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18353062 Supplier Novus Biologicals Supplier No. NBP316834100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

RNF40 Monoclonal antibody specifically detects RNF40 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF40
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8I7R5
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias 95 kDa retinoblastoma-associated protein, BRE1-B, BRE1BMGC13051, EC 6.3.2,95 kDa retinoblastoma protein binding protein, EC 6.3.2.-, KIAA0661BRE1 E3 ubiquitin ligase homolog B, Rb-associated protein, RBP95E3 ubiquitin-protein ligase BRE1B, ring finger protein 40DKFZp686K191, STARING
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 231-431 of human RNF40 (O75150).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Chromatin Research, Vision, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 9810
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.