missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF40 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38267-20ul
This item is not returnable.
View return policy
Description
RNF40 Polyclonal antibody specifically detects RNF40 in Human samples. It is validated for ELISA,Western Blot
Specifications
| RNF40 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| 95 kDa retinoblastoma-associated protein, BRE1-B, BRE1BMGC13051, EC 6.3.2,95 kDa retinoblastoma protein binding protein, EC 6.3.2.-, KIAA0661BRE1 E3 ubiquitin ligase homolog B, Rb-associated protein, RBP95E3 ubiquitin-protein ligase BRE1B, ring finger protein 40DKFZp686K191, STARING | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human RNF40 (NP_055586.1).,, Sequence:, MSGPGNKRAAGDGGSGPPEKKLSREEKTTTTLIEPIRLGGISSTEEMDLKVLQFKNKKLAERLEQRQACEDELRERIEKLEKRQATDDATLLIVNRYWAQLDETVEALLRCHESQGELSSAPEAPGTQEGPTCDGTPLPEPGTSELRDPLLMQLRPPLSEPALAFVVALGASSSEEVELELQGRMEFSKAAVSRVVEASDRLQRRVEELCQRVYSRGDSEPLSEAAQAHTRELGRENRRLQDLATQLQEKHHRISLEYSELQDKVTSAETKVLEMETTVEDLQWDIEKLRKREQKLNKHL | |
| 20 μL | |
| Chromatin Research, Vision, Zinc Finger | |
| 9810 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction