missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF4 Monoclonal Antibody (OTI9F10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527424
This item is not returnable.
View return policy
Description
RNF4 Monoclonal Antibody for Western Blot
The protein encoded by this gene contains a RING finger motif and acts as a transcription regulator. This protein has been shown to interact with, and inhibit the activity of, TRPS1, a transcription suppressor of GATA-mediated transcription. Transcription repressor ZNF278/PATZ is found to interact with this protein, and thus reduce the enhancement of androgen receptor-dependent transcription mediated by this protein. Studies of the mouse and rat counterparts suggested a role of this protein in spermatogenesis. A pseudogene of this gene is found on chromosome 1.
Specifications
| RNF4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O88846, P78317, Q9QZS2 | |
| RNF4 | |
| Human recombinant protein fragment corresponding to amino acids 1-190 of RNF4 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI9F10 | |
| Unconjugated | |
| RNF4 | |
| AU018689; E3 ubiquitin ligase RNF4; E3 ubiquitin-protein ligase RNF4; Gtrgeo8; Protein SNURF; RES4-26; RING finger protein 4; RING-type E3 ubiquitin transferase RNF4; RNF4; SLX5; small nuclear RING finger protein; SNURF | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 19822, 29274, 6047 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction