missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF36 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38213-100ul
This item is not returnable.
View return policy
Description
RNF36 Polyclonal antibody specifically detects RNF36 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| RNF36 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| RFP-like domain-containing protein trimless, RING finger protein 36HSD34, RNF36, Trif, trimless, tripartite motif containing 69, tripartite motif protein 69, tripartite motif-containing 69, tripartite motif-containing protein 69 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-341 of human RNF36 (NP_542783.2).,, Sequence:, MEEELAIQQGQLETTLKELQTLRNMQKEAIAAHKENKLHLQQHVSMEFLKLHQFLHSKEKDILTELREEGKALNEEMELNLSQLQEQCLLAKDMLVSIQAKTEQQNSFDFLKDITTLLHSLEQGMKVLATRELISRKLNLGQYKGPIQYMVWREMQDTLCPGLSPLTLDPKTAHPNLVLSKSQTSVWHGDIKKIMPDDPERFDSSVAVLGSRGFTSGKWYWEVEVAKKTKWTVGVVRESIIRKGSCPLTPEQGFWLLRLRNQTDLKALDLPSFSLTLTNNLDKVGIYLDYEGGQLSFYNAKTMTHIYTFSNTFMEKLYPYFCPCLNDGGENKEPLHILHPQ | |
| 100 μL | |
| Zinc Finger | |
| 140691 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction