missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF36 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20188
This item is not returnable.
View return policy
Description
RNF36 Polyclonal specifically detects RNF36 in Human samples. It is validated for Western Blot.
Specifications
| RNF36 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| RFP-like domain-containing protein trimless, RING finger protein 36HSD34, RNF36, Trif, trimless, tripartite motif containing 69, tripartite motif protein 69, tripartite motif-containing 69, tripartite motif-containing protein 69 | |
| Rabbit | |
| 57 kDa | |
| 0.1 mL | |
| Zinc Finger | |
| 140691 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q86WT6 | |
| TRIM69 | |
| Recombinant protein encompassing a sequence within the center region of human RNF36. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction