missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF34 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10629-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene contains a RINF finger, a motif known to be involved in protein-protein and protein-DNA interactions. This protein interacts with DNAJA3/hTid-1, which is a DnaJ protein reported to function as a modulator of apoptosis. Overexpression of this gene in Hela cells was shown to confer the resistance to TNF-alpha induced apoptosis, suggesting an anti-apoptotic function of this protein. This protein can be cleaved by caspase-3 during the induction of apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been reported.Specifications
RNF34 | |
Polyclonal | |
Unconjugated | |
RNF34 | |
CARP 1, Caspase regulator CARP1, FYVE RING finger protein Momo, hRFI, RFI, RIF, RIFF, ring finger protein 34, RING finger protein RIFF, RNF34 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
282845, 80196, 80751 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6AYH3, Q969K3, Q99KR6 | |
Rnf34 | |
RNF34 Fusion Protein Ag0982 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |