missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF34 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567332
This item is not returnable.
View return policy
Description
Immunogen sequence: NIVCKACGLSF SVFRKKHVCC DCKKDFCSVC SVLQENLRRC STCHLLQETA FQRPQLMRLK VKDL Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.
The protein encoded by this gene contains a RINF finger, a motif known to be involved in protein-protein and protein-DNA interactions. This protein interacts with DNAJA3/hTid-1, which is a DnaJ protein reported to function as a modulator of apoptosis. Overexpression of this gene in Hela cells was shown to confer the resistance to TNF-alpha induced apoptosis, suggesting an anti-apoptotic function of this protein. This protein can be cleaved by caspase-3 during the induction of apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
| RNF34 | |
| Polyclonal | |
| Unconjugated | |
| RNF34 | |
| AW061037; AW536122; BC004042; C88279; CARP1; CARP-1; Caspase regulator CARP1; caspases-8 and -10-associated RING finger protein 1; E3 ubiquitin-protein ligase RNF34; FLJ21786; FYVE-RING finger protein MOMO; hRFI; Human RING finger homologous to inhibitor of apoptosis protein; Momo; phafin 1; phafin-1; RFI; RIF; RIFF; ring finger protein 34; ring finger protein 34, E3 ubiquitin protein ligase; RING finger protein MOMO; RING finger protein RIFF; RING-type E3 ubiquitin transferase RNF34; RNF34 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80196 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q969K3 | |
| RNF34 | |
| Recombinant Human RNF34. Recombinant protein control fragment (Product #RP-105587). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction