missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF31 Recombinant Rabbit Monoclonal Antibody (HL2295)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550479
This item is not returnable.
View return policy
Description
RNF31 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Lyn proto oncogene (Lcl/Yes related novel protein tyrosine kinase) is encoded by the Lyn gene located on chromosome 8 in humans and belongs to the Src family kinase. It is a non-receptor tyrosine kinase that acts as a mediator in transducing cellular signaling. The most well studied expression of Lyn is in hematopoietic cell types, both lymphoid and myeloid cells with exclusion in T lymphocytes. It has also been observed that Lyn plays diverse roles such as in regulation of B cell receptor signaling, mast cell signaling and Epithelial - Mesenchymal Transition (EMT).
Specifications
| RNF31 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with no preservative | |
| Q924T7, Q96EP0 | |
| RNF31 | |
| Recombinant fragmemt of human RNF31. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL2295 | |
| Unconjugated | |
| RNF31 | |
| AL033293; BC031509; E3 ubiquitin-protein ligase RNF31; Flj10111; FLJ23501; HOIL-1-interacting protein; HOIP; mFLJ00217; MGC19975; Paul; putative Ariadne-like ubiquitin ligase; RING finger protein 31; RING-type E3 ubiquitin transferase RNF31; Rnf31; ZIBRA; Zinc in-between-RING-finger ubiquitin-associated domain protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 268749, 364386, 55072 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction