missing translation for 'onlineSavingsMsg'
Learn More

RNF26 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18409137 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409137 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409137 Supplier Novus Biologicals Supplier No. NBP281997AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RNF26 Polyclonal antibody specifically detects RNF26 in Human, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF26
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias MGC2642, ring finger protein 26, ring finger protein with leucine zipper
Host Species Rabbit
Immunogen RNF26 antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of human RNF26. Amino Acid Squence: RGRERLNEEEPPGGQDPWK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 79102
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.