missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNF23 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15790112 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15790112 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15790112 Supplier Invitrogen™ Supplier No. PA562321

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: GSMVEIAKQL QAVKRKIRDE SLCPQHHEAL SLFCYEDQEA VCLICAISHT HRAHTVVPLD DATQEYKE Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The function of this protein has not been identified. This gene lies within the major histocompatibility complex class I region on chromosome 6. Alternate splicing results in two transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF23
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIM39
Gene Accession No. Q9HCM9
Gene Alias 1100001D15Rik; E130103K13Rik; E3 ubiquitin-protein ligase TRIM39; mKIAA4179; RBCC-B30.2; ring finger protein 23; RING-B box-coiled coil-B30.2; RING-B box-coiled-coil-B30.2; RING-type E3 ubiquitin transferase TRIM39; Rnf23; Testis-abundant finger protein; tfp; TRIM39; TRIM39B; tripartite motif containing 39; tripartite motif protein 39; tripartite motif-containing 39; tripartite motif-containing protein 39
Gene Symbols TRIM39
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNF23. Recombinant protein control fragment (Product #RP-99607).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56658
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.