missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF23 Antibody (OTI2A6), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73926IR
This item is not returnable.
View return policy
Description
RNF23 Monoclonal specifically detects RNF23 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot.
Specifications
| RNF23 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TRIM39 | |
| Recombinant protein fragment corresponding to amino acids 144-378 of human RNF23 (NP_742013) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG1 |
| Western Blot | |
| OTI2A6 | |
| Western Blot | |
| MGC32984, RING finger protein 23RNF23TFPtripartite motif-containing 39, Testis-abundant finger protein, TRIM39B, tripartite motif containing 39, tripartite motif-containing protein 39 | |
| Mouse | |
| Affinity Purified | |
| Zinc Finger | |
| 56658.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction