missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF220 Polyclonal Antibody

RNF220 Polyclonal Antibody
Brand: Invitrogen PA5110308
This item is not returnable.
View return policy
Description
RNF220 Polyclonal Antibody for Western Blot
Specifications
| RNF220 | |
| Polyclonal | |
| Unconjugated | |
| RNF220 | |
| 4732477A13; 4931406I20Rik; 5730503K05Rik; AV302541; C1orf164; E3 ubiquitin-protein ligase RNF220; ring finger protein 220; RING-type E3 ubiquitin transferase Rnf220; RNF220 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 500532, 55182, 66743 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.86 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q5VTB9, Q6PDX6 | |
| RNF220 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 267-566 of human RNF220 (NP_060620.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction