Learn More
RNF219 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24988-1-AP-150UL
 View more versions of this product
Product Code. 16873505
  • £349.00 / 150µL
Description
RNF219 (Ring Finger Protein 219, C13orf7) is a Protein Coding gene.
Specifications
Specifications
RNF219
Polyclonal
Unconjugated
RNF219
C13orf7, ring finger protein 219, RNF219
Rabbit
Antigen Affinity Chromatography
RUO
79596
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.48 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5W0B1
RNF219
RNF219 Fusion Protein Ag21767
150 μL