missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF213 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518650
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.
RNF213 is an E3 ubiquitin-protein ligase involved in angiogenesis. Involved in the non-canonical Wnt signaling pathway in vascular development: acts by mediating ubiquitination and degradation of FLNA and NFATC2 downstream of RSPO3, leading to inhibit the non-canonical Wnt signaling pathway and promoting vessel regression. Also has ATPase activity.
Specifications
| RNF213 | |
| Polyclonal | |
| Unconjugated | |
| RNF213 | |
| 6030403J01; ALK lymphoma oligomerization partner on chromosome 17; ALO17; C17orf27; D11Ertd759e; E3 ubiquitin-protein ligase RNF213; KIAA1554; KIAA1618; mKIAA1554; MYMY2; Mysterin; MYSTR; NET57; protein ALO17; RGD1308168; RING finger protein 213; RING-type E3 ubiquitin transferase RNF213; RNF213 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 57674 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q63HN8 | |
| RNF213 | |
| Synthetic peptide sequence (KTAAVLKWNRE) corresponding to the C-terminus amino acids of RNF213 (aa 5195-5206). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction