missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF213 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555418
This item is not returnable.
View return policy
Description
Immunogen sequence: KDPVCLPCDH VHCLRCLRAW FASEQMICPY CLTALPDEFS PAVSQAHREA IEKHARFRQM CNSFFVDLVS TICFKDNAPP EKEVIESLLS LLFVQKGRLR DAAQRHCEHT KSLSPFNDVV DKTPVIRSVI LKLLLKYS Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 79%.
RNF213 is an E3 ubiquitin-protein ligase involved in angiogenesis. Involved in the non-canonical Wnt signaling pathway in vascular development: acts by mediating ubiquitination and degradation of FLNA and NFATC2 downstream of RSPO3, leading to inhibit the non-canonical Wnt signaling pathway and promoting vessel regression. Also has ATPase activity.
Specifications
| RNF213 | |
| Polyclonal | |
| Unconjugated | |
| RNF213 | |
| 6030403J01; ALK lymphoma oligomerization partner on chromosome 17; ALO17; C17orf27; D11Ertd759e; E3 ubiquitin-protein ligase RNF213; KIAA1554; KIAA1618; mKIAA1554; MYMY2; Mysterin; MYSTR; NET57; protein ALO17; RGD1308168; RING finger protein 213; RING-type E3 ubiquitin transferase RNF213; RNF213 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57674 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q63HN8 | |
| RNF213 | |
| Recombinant protein corresponding to Human RNF213. Recombinant protein control fragment (Product #RP-89516). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction