missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RNF213 Monoclonal antibody specifically detects RNF213 in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | RNF213 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 5C12 |
| Conjugate | Unconjugated |
| Dilution | Western Blot, ELISA |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Alias | ALK lymphoma oligomerization partner on chromosome 17, C17orf27, chromosome 17 open reading frame 27, DKFZp762N1115, FLJ13051, KIAA1554MGC46622, KIAA1618, MGC9929, NET57, protein ALO17, ring finger protein 213 |
| Host Species | Mouse |
| Immunogen | C17orf27 (NP_065965, 2016 a.a. ∽ 2112 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LSPENAKLLSTFLNQTGLDAFLLELHEMIILKLKNPQTQTEERFRPQWSLRDTLVSYMQTKESEILPEMASQFPEEILLASCVSVWKTAAVLKWNRE |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?