missing translation for 'onlineSavingsMsg'
Learn More

RNF2 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18319760 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18319760 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18319760 Supplier Novus Biologicals Supplier No. NBP177826JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

RNF2 Polyclonal specifically detects RNF2 in Human, Mouse, Chinese Hamster samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF2
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias BAP-1, BAP1RING finger protein 1B, DINGRING finger protein BAP-1, EC 6.3.2, HIPI3RING1BE3 ubiquitin-protein ligase RING2, Huntingtin-interacting protein 2-interacting protein 3, Protein DinG, ring finger protein 2HIP2-interacting protein 3, RING1b, RING2EC 6.3.2.-
Host Species Goat
Immunogen RNF2 Antibody was prepared from whole goat serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region near aa 180-205 of human RNF2 protein. (Uniprot: Q99496)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Signal Transduction, Transcription Factors and Regulators, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 6045
Target Species Human, Mouse, Chinese Hamster
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.