missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF19A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554861
This item is not returnable.
View return policy
Description
Immunogen sequence: EMCTDKNSIF STNTSSDNGL TSISKQIGDF IECPLCLLRH SKDRFPDIMT CHHRSCVDCL Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 93%.
This gene encodes a member of the ring between ring fingers (RBR) protein family, and the encoded protein contains two RING-finger motifs and an in between RING fingers motif. This protein is an E3 ubiquitin ligase that is localized to Lewy bodies, and ubiquitylates synphilin-1, which is an interacting protein of alpha synuclein in neurons. The encoded protein may be involved in amyotrophic lateral sclerosis and Parkinson's disease. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Jul 2013].
Specifications
| RNF19A | |
| Polyclonal | |
| Unconjugated | |
| RNF19A | |
| AA032313; Dorfin; Double ring-finger protein; E3 ubiquitin-protein ligase RNF19A; Gametogenesis-expressed protein GEG-154; Geg-154; p38; protein p38 interacting with transcription factor Sp1; ring finger protein (C3HC4 type) 19; ring finger protein 19; ring finger protein 19A; ring finger protein 19A, E3 ubiquitin protein ligase; ring finger protein 19A, RBR E3 ubiquitin protein ligase; ring-IBR-ring domain containing protein Dorfin; Rnf19; RNF19A; Ubce7ip2; UBCM4-interacting protein 117; ubiquitin conjugating enzyme 7 interacting protein 2; UIP117; XY body protein; XYbp | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25897 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NV58 | |
| RNF19A | |
| Recombinant protein corresponding to Human RNF19A. Recombinant protein control fragment (Product #RP-93729). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction