missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF186 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579928
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human SW620 cell, human HepG2 cell.
The RNF186 gene encodes the RING finger protein 186, a member of the RING-type E3 ubiquitin ligase family. RNF186 is involved in the ubiquitin-proteasome pathway, playing a crucial role in tagging target proteins for degradation, thus influencing protein turnover and cellular homeostasis. This process is essential for regulating various cellular functions, including signal transduction, apoptosis, and response to stress. RNF186 has been implicated in the unfolded protein response (UPR) within the endoplasmic reticulum, where it helps in managing protein folding and maintaining cellular homeostasis, particularly under conditions of endoplasmic reticulum stress. Alterations in RNF186 expression or function have been associated with disorders such as inflammatory bowel disease and ulcerative colitis, suggesting its role in gastrointestinal health and disease. Research into RNF186 focuses on its regulatory mechanisms within the ubiquitin-proteasome system and its potential as a therapeutic target for conditions associated with protein misfolding and cellular stress responses.
Specifications
| RNF186 | |
| Polyclonal | |
| Unconjugated | |
| RNF186 | |
| 9130020G10Rik; E3 ubiquitin-protein ligase RNF186; RING finger protein 186; RNF186 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54546 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| Q9NXI6 | |
| RNF186 | |
| E. coli-derived human RNF186 recombinant protein (Position: R86-L159). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction