missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF185 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578615
This item is not returnable.
View return policy
Description
Positive Control: human RNF185-transfected 293T Predicted Reactivity: Mouse (98%), Rat (98%), Zebrafish (83%), Xenopus laevis (91%), Chicken (87%), Bovine (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RNF185 is a E3 ubiquitin-protein ligase that regulates selective mitochondrial autophagy by mediating Lys-63-linked polyubiquitination of BNIP1. RNF185 acts in the endoplasmic reticulum (ER)-associated degradation (ERAD) pathway, which targets misfolded proteins that accumulate in the endoplasmic reticulum (ER) for ubiquitination and subsequent proteasome-mediated degradation. RNF185 protects cells from ER stress-induced apoptosis. is responsible for the cotranslational ubiquitination and degradation of CFTR in the ERAD pathway, and is preferentially associates with the E2 enzymes UBE2J1 and UBE2J2.
Specifications
| RNF185 | |
| Polyclonal | |
| Unconjugated | |
| RNF185 | |
| BSK65-MONO1; BSK65-MONO2; BSK65-PANC1; BSK65-PANC2; BSK65-TEST1; BSK65-TEST2; BSK65-TEST3; E3 ubiquitin-protein ligase RNF185; FLJ38628; I79_017427; ring finger protein 185; RING-type E3 ubiquitin transferase RNF185; rn185; Rnf185 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 91445 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.68 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q96GF1 | |
| RNF185 | |
| Recombinant protein encompassing a sequence within the center region of human RNF185. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction