missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF183 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | RNF183 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
RNF183 Polyclonal specifically detects RNF183 in Mouse samples. It is validated for Western Blot.Specifications
| RNF183 | |
| Polyclonal | |
| Rabbit | |
| Q8QZS5 | |
| 138065 | |
| Synthetic peptides corresponding to the N terminal of Rnf183. Immunizing peptide sequence LSLVTPARRRLLCPLCRQPTVLASGQPVTDLPTDTAMLTLLRLEPHHVIL. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| FLJ31197, MGC4734, ring finger protein 183 | |
| RNF183 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title