missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF181 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20408-1-AP-20UL
This item is not returnable.
View return policy
Description
RNF181 binds the integrin alpha-IIb /beta-3 complex and has E3 ubiquitin ligase activity (Brophy et al., 2008 ).Specifications
| RNF181 | |
| Polyclonal | |
| Unconjugated | |
| RNF181 | |
| HSPC238, ring finger protein 181, RN181, RNF181 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 297337, 51255, 66510 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Western Blot | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6AXU4, Q9CY62, Q9P0P0 | |
| RNF181 | |
| RNF181 Fusion Protein Ag14233 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction