missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF170 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562270
This item is not returnable.
View return policy
Description
Immunogen sequence: CGACIIAYWR YGSWLGAISC PICRQTVTLL LTVFGEDDQS QDVLRLHQDI NDYNRRFSGQ PRSIMERIMD LPT Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
RNF170 is a multi-pass membrane protein. It contains 1 RING-type zinc finger. The exact function of RNF170 remains unknown.
Specifications
| RNF170 | |
| Polyclonal | |
| Unconjugated | |
| RNF170 | |
| 6720407G21Rik; ADSA; AI481227; E3 ubiquitin-protein ligase RNF170; putative LAG1-interacting protein; RING finger protein 170; RING finger protein 170; E3 ubiquitin-protein ligase RNF170; RING-type E3 ubiquitin transferase RNF170; Rnf170; SNAX1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 81790 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96K19 | |
| RNF170 | |
| Recombinant protein corresponding to Human RNF170. Recombinant protein control fragment (Product #RP-101241). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction