missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ RNF169 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-28892
This item is not returnable.
View return policy
Description
RNF169 Polyclonal antibody specifically detects RNF169 in Human samples. It is validated for Immunoprecipitation
Specifications
| RNF169 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| Unconjugated | |
| EC 3.4.24.57, KIAA1991EC 3.1.4.16, ring finger protein 169 | |
| Between 200 and 250 | |
| 100 μg | |
| Zinc Finger | |
| 254225 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction